Skip to Content
Merck
2-Methoxy-3-(1-methylpropyl)pyrazine ≥98%, FG

2-Methoxy-3-(1-methylpropyl)pyrazine

2-Methoxy-3-(1-methylpropyl)pyrazine ≥98%, FG
Synonym(s):
2-sec-Butyl-3-methoxypyrazine
Empirical Formula (Hill Notation):
C9H14N2O
CAS Number:
Molecular Weight:
166.22
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing