Skip to Content
Merck
2-Ethyl-1-hexanol analytical standard

2-Ethyl-1-hexanol

2-Ethyl-1-hexanol analytical standard
Synonym(s):
Isooctyl alcohol, 2-Ethyl-1-hexanol
Linear Formula:
CH3(CH2)3CH(C2H5)CH2OH
CAS Number:
Molecular Weight:
130.23
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5