![2-Ethoxyethanol (stabilised) for synthesis](/deepweb/assets/sigmaaldrich/product/structures/155/878/9466679f-ea2d-4c59-a673-32e59e4bbd21/640/9466679f-ea2d-4c59-a673-32e59e4bbd21.png)
2-Ethoxyethanol (stabilised)
![2-Ethoxyethanol (stabilised) for synthesis](/deepweb/assets/sigmaaldrich/product/structures/155/878/9466679f-ea2d-4c59-a673-32e59e4bbd21/640/9466679f-ea2d-4c59-a673-32e59e4bbd21.png)
Synonym(s):
2-Ethoxyethanol, Ethyl glycol, Ethylene glycol monoethyl ether
Linear Formula:
C2H5OCH2CH2OH
CAS Number:
Molecular Weight:
90.12
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing