Skip to Content
Merck
2-Cyclopentene-1-acetic acid 98%

2-Cyclopentene-1-acetic acid

2-Cyclopentene-1-acetic acid 98%
Synonym(s):
2-Cyclopenten-1-ylacetic acid
Linear Formula:
C5H7CH2CO2H
CAS Number:
Molecular Weight:
126.15
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing