Skip to Content
Merck
2-Chloromethyl-2,3-dihydrothieno[3,4-b]-1,4-dioxine 95%

2-Chloromethyl-2,3-dihydrothieno[3,4-b]-1,4-dioxine

2-Chloromethyl-2,3-dihydrothieno[3,4-b]-1,4-dioxine 95%
Empirical Formula (Hill Notation):
C7H7ClO2S
CAS Number:
Molecular Weight:
190.65

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing