Skip to Content
Merck
2-(Aminomethyl)piperidine 97%

2-(Aminomethyl)piperidine

2-(Aminomethyl)piperidine 97%
Synonym(s):
(RS)-1-(Piperidin-2-yl)methanamine, 2-Piperidinemethanamine
Empirical Formula (Hill Notation):
C6H14N2
CAS Number:
Molecular Weight:
114.19
EC Number:

Products

Product Category
Brand
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing