2-Acetoxy-3-butanone
Synonym(s):
Acetoin acetate
Empirical Formula (Hill Notation):
C6H10O3
CAS Number:
Molecular Weight:
130.14
Products
Product Category
Brand
Formula Weight
Purity
Agency
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing