25-Hydroxycholesterol
Synonym(s):
5-Cholestene-3β,25-diol
Empirical Formula (Hill Notation):
C27H46O2
CAS Number:
Molecular Weight:
402.65
Products
Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Physical Form
Purity
Showing 1-2 of 2
Compare
Product Number
Description
Pricing