Skip to Content
Merck
2-(4-Methyl-1-piperidinyl)ethanamine

2-(4-Methyl-1-piperidinyl)ethanamine

2-(4-Methyl-1-piperidinyl)ethanamine
Empirical Formula (Hill Notation):
C8H18N2
CAS Number:
Molecular Weight:
142.24

Products

Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing