2,3-Dimethyl-2,3-Butanediamine Dihydrochloride
Synonym(s):
2,3-Dimethyl-2,3-butanediamine dihydrochloride, 1,1,2,2-Tetramethylethylenediamine dihydrochloride
Empirical Formula (Hill Notation):
C6H16N2 · 2HCl
CAS Number:
Molecular Weight:
189.13
Products
Brand
Formula Weight
Manufacturer
Product Line
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2