2-(2H-Benzotriazol-2-yl)-4-methyl-6-(2-propenyl)phenol
Synonym(s):
2-(3-Allyl-2-hydroxy-5-methylphenyl)-2H-benzotriazole, 2-Allyl-6-(2H-benzotriazol-2-yl)-p-cresol
Empirical Formula (Hill Notation):
C16H15N3O
CAS Number:
Molecular Weight:
265.31
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Markush Class
Markush Group
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing