![2,2′-Dibromobiphenyl 97%](/deepweb/assets/sigmaaldrich/product/structures/651/380/54252fd2-0e8c-48c6-acfe-b7ae4fff97cf/640/54252fd2-0e8c-48c6-acfe-b7ae4fff97cf.png)
2,2′-Dibromobiphenyl
![2,2′-Dibromobiphenyl 97%](/deepweb/assets/sigmaaldrich/product/structures/651/380/54252fd2-0e8c-48c6-acfe-b7ae4fff97cf/640/54252fd2-0e8c-48c6-acfe-b7ae4fff97cf.png)
Empirical Formula (Hill Notation):
C12H8Br2
CAS Number:
Molecular Weight:
312.00
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing