2,2′-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt
Synonym(s):
AzBTS-(NH4)2, Diammonium 2,2′-azino-bis(3-ethylbenzothiazoline-6-sulfonate)
Empirical Formula (Hill Notation):
C18H24N6O6S4
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing