Skip to Content
Merck
1-(Triphenylmethyl)imidazole 97%

1-(Triphenylmethyl)imidazole

1-(Triphenylmethyl)imidazole 97%
Empirical Formula (Hill Notation):
C22H18N2
CAS Number:
Molecular Weight:
310.39

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing