Skip to Content
Merck
(1S)-(+)-Menthyl chloroformate optical purity ee: 97% (GLC)

(1S)-(+)-Menthyl chloroformate

(1S)-(+)-Menthyl chloroformate optical purity ee: 97% (GLC)
Empirical Formula (Hill Notation):
C11H19ClO2
CAS Number:
Molecular Weight:
218.72

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing