Skip to Content
Merck
(1S,2R)-(+)-Ephedrine hydrochloride 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

(1S,2R)-(+)-Ephedrine hydrochloride

(1S,2R)-(+)-Ephedrine hydrochloride 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Synonym(s):
(1S,2R)-(+)-α-(1-Methylaminoethyl)benzyl alcohol hydrochloride, (1S,2R)-(+)-2-Methylamino-1-phenyl-1-propanol hydrochloride, D-α-(1-Methylaminoethyl)benzyl alcohol hydrochloride
Linear Formula:
C6H5CH[CH(NHCH3)CH3]OH · HCl
CAS Number:
Molecular Weight:
201.69
EC Number:

Products

Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Format
Application
Markush Class
Markush Group
Optical Isomer
Package Size
Reference Material Type
Special Grade
Technique