Skip to Content
Merck
vibrant-m

1H,1H,2H,2H-Perfluorodecanesulfonic acid

vibrant-m
Synonym(s):
1H,1H′,2H,2H′-Perfluorodecane sulfonic acid, 8:2 Fluorotelomer sulfonic acid
Empirical Formula (Hill Notation):
C10H5F17O3S
CAS Number:
Molecular Weight:
528.18

Products

Brand
Formula Weight
Manufacturer
Quality Segment
Reference Material Type
Special Grade