Skip to Content
Merck
1-Ethynyl-2,4-difluorobenzene 97%

1-Ethynyl-2,4-difluorobenzene

1-Ethynyl-2,4-difluorobenzene 97%
Linear Formula:
HC≡CC6H3F2
CAS Number:
Molecular Weight:
138.11

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing