Skip to Content
Merck
1-Aminobenzotriazole

1-Aminobenzotriazole

1-Aminobenzotriazole
Synonym(s):
1-Benzotriazolamine, ABT
Empirical Formula (Hill Notation):
C6H6N4
CAS Number:
Molecular Weight:
134.14

Products

Product Category
Brand
Sustainability
Feature
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing