![1,4-Dichlorobenzene-d4 solution certified reference material, 2000 μg/mL in methanol](/deepweb/assets/sigmaaldrich/product/images/222/740/35b14787-a8c2-4c84-95f0-46274ec15db7/640/35b14787-a8c2-4c84-95f0-46274ec15db7.jpg)
1,4-Dichlorobenzene-d4 solution
![1,4-Dichlorobenzene-d4 solution certified reference material, 2000 μg/mL in methanol](/deepweb/assets/sigmaaldrich/product/images/222/740/35b14787-a8c2-4c84-95f0-46274ec15db7/640/35b14787-a8c2-4c84-95f0-46274ec15db7.jpg)
Empirical Formula (Hill Notation):
C6D4Cl2
CAS Number:
Molecular Weight:
151.03
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing