Skip to Content
Merck
1-[3-(Trimethoxysilyl)propyl]urea 97%

1-[3-(Trimethoxysilyl)propyl]urea

1-[3-(Trimethoxysilyl)propyl]urea 97%
Linear Formula:
(CH3O)3Si(CH2)3NHCONH2
CAS Number:
Molecular Weight:
222.31

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing