Skip to Content
Merck
1,2-Dipalmitoyl-sn-glycero-3-phosphocholine semisynthetic, ≥99%

1,2-Dipalmitoyl-sn-glycero-3-phosphocholine

1,2-Dipalmitoyl-sn-glycero-3-phosphocholine semisynthetic, ≥99%
Synonym(s):
(7R)-4-Hydroxy-N,N,N-trimethyl-10-oxo-7-[(1-oxohexadecyl)oxy]- 3,5,9-trioxa-4-phosphapentacosan-1-aminium 4-oxide, inner salt, 1,2-Dihexadecanoyl-sn-glycero-3-phosphocholine, 1,2-Dihexadecanoyl-sn-glycero-3-phosphorylcholine, 1,2-Dipalmitoyl-sn-glycero-3-phosphorylcholine, 1,2-Dipalmitoyl-L-lecithin, L-α-Phosphatidylcholine, dipalmitoyl, L-β,γ-Dipalmitoyl-α-lecithin, DPPC, PC(16:0/16:0)
Empirical Formula (Hill Notation):
C40H80NO8P
CAS Number:
Molecular Weight:
734.04
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Functional Group
Optical Isomer
Showing 1-2 of 2