
1,2-Dichloroethane-d4

Synonym(s):
Ethylene-d4 dichloride
Linear Formula:
ClCD2CD2Cl
CAS Number:
Molecular Weight:
102.98
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Isotope
Application
Technique
Showing 1-1 of 1