Skip to Content
Merck
1,2,3,4-Tetrahydro-2-naphthoic acid 98%

1,2,3,4-Tetrahydro-2-naphthoic acid

1,2,3,4-Tetrahydro-2-naphthoic acid 98%
Synonym(s):
Tetralin-2-carboxylic acid
Empirical Formula (Hill Notation):
C11H12O2
CAS Number:
Molecular Weight:
176.21
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing