Product Name
7-(Diethylamino)coumarin-3-carbohydrazide BioReagent, suitable for fluorescence, ≥95% (HPCE)
Product Number
36798
Product Brand
sigma
CAS Number
100343-98-4
Molecular Weight
275.30
Test
Specification
APPEARANCE (COLOR)Yellow to Dark Yellow
APPEARANCE (FORM)Powder or Crystals
PURITY (HPCE AREA %)≥ 95.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EMISSION MAXIMUM463 - 473 nm
REMARKS ON LUMINESCENCEEXC. 450 NM IN METHANOL