Product Name
Methyl benzo[b]thiophene-2-carboxylate 97%
Product Number
666432
Product Brand
aldrich
CAS Number
22913-24-2
Molecular Weight
192.23
Test | Specification |
---|---|
APPEARANCE | WHITE TO YELLOW POWDER, CRYSTALS OR |
CRYSTALLINE POWDER AND/OR CHUNKS | |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
GAS LIQUID | 96.5% (MINIMUM) |
REVISED SEPTEMBER 19, 2006 RJM |