Merck
Nickel Catalysts
Periodic Table Symbol for Nickel
Nickel catalysis plays a central role in many synthetic transformations ranging from cross coupling reactions in which carbon–carbon bonds are formed, to the reduction of electron-rich carbon bonds with raney nickel catalysts. These nickel catalysts span a range of oxidation states: Nickel (0), nickel (II), nickel (III) and nickel (IV). The types of Ni catalysts available for immediate purchase are aluminum nickel (Al Ni) alloys, ammonium nickel hydrates, Ni COD, Ni halides (chlorides, bromides, fluorides and iodides), Ni cyclopentadienyls, nickel metal, nickel acacs, and Raney Nickel catalysts that are products of W.R. Grace and Company.
Products
Sort by Relevance
Showing 1-20 of 87
Change View
Showing 1-20 of 87
Compare
Product Number
Product Name
Product Description
Pricing
You have viewed 1-20 of 87 results