Skip to Content
Merck
All Photos(3)

Key Documents

V900488

Sigma-Aldrich

Cholic acid

Vetec, reagent grade, 98%

Synonym(s):

3α,7α,12α-Trihydroxy-5β-cholanic acid, Cholanic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H40O5
CAS Number:
Molecular Weight:
408.57
Beilstein:
2822009
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

biological source

bovine bile

grade

reagent grade

description

non-denaturing ionic detergent

product line

Vetec

Assay

98%

form

powder

mol wt

408.57 g/mol

mp

200-201 °C (lit.)

functional group

carboxylic acid

shipped in

ambient

storage temp.

room temp

SMILES string

[H][C@@]12C[C@H](O)CC[C@]1(C)[C@@]3([H])C[C@H](O)[C@]4(C)[C@]([H])(CC[C@@]4([H])[C@]3([H])[C@H](O)C2)[C@H](C)CCC(O)=O

InChI

1S/C24H40O5/c1-13(4-7-21(28)29)16-5-6-17-22-18(12-20(27)24(16,17)3)23(2)9-8-15(25)10-14(23)11-19(22)26/h13-20,22,25-27H,4-12H2,1-3H3,(H,28,29)/t13-,14+,15-,16-,17+,18+,19-,20+,22+,23+,24-/m1/s1

InChI key

BHQCQFFYRZLCQQ-OELDTZBJSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Non-denaturing ionic detergent used for extraction of membrane proteins.

Biochem/physiol Actions

Bile Acid

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zheng Pan et al.
Carbohydrate polymers, 94(1), 394-399 (2013-04-03)
Self-assembled amphiphilic N-(2,3-dihydroxypropyl)-chitosan-cholic acid (DHP-CS-CHO) micelle was prepared as a carrier for paclitaxel. DHP-CS-CHO was synthesized by grafted small molecules cholic acid and glycidol onto primary amine group of chitosan, respectively. The DHP-CS-CHO formed uniform micelles (size=212.4±3.1 nm) with a
Yu Shao et al.
ACS applied materials & interfaces, 6(13), 10381-10392 (2014-06-13)
To develop a feasible and efficient nanocarrier for potential clinical application, a series of photo and redox dual responsive reversibly cross-linked micelles have been developed for the targeted anticancer drug delivery. The nanocarrier can be cross-linked efficiently via a clean
Sylwia Mildner-Szkudlarz et al.
Journal of the science of food and agriculture, 93(13), 3271-3278 (2013-04-16)
New breads fortified with two different forms of grape by-products, namely dried powdered skins (PGP) and freeze-dried extract therefrom (EGP), were characterised and their protective effect against hypercholesterolaemia in rats was studied. The phenolic compound profiles of supplemented breads were
Florence Janvier et al.
Journal of the mechanical behavior of biomedical materials, 18, 100-107 (2012-12-25)
Elevated red blood cell (RBC) aggregation increases low-shear blood viscosity and is closely related to several pathophysiological diseases such as atherosclerosis, thrombosis, diabetes, hypertension, cancer, and hereditary chronic hemolytic conditions. Non-ionic linear polymers such as poly(ethylene glycol) (PEG) and Pluronic
Chiara Gabbi et al.
Digestive and liver disease : official journal of the Italian Society of Gastroenterology and the Italian Association for the Study of the Liver, 44(12), 1018-1026 (2012-08-14)
Non-alcoholic fatty liver disease, one of the most prevalent liver disorders in Western countries, is characterized by hepatic accumulation of triglycerides. Bile acids have long been known to affect triglyceride homeostasis through a not completely understood mechanism. To analyse the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service