Skip to Content
Merck
All Photos(1)

Key Documents

SY3H6E4112FE

Synthonix Corporation

(3-Cyano-4-hydroxyphenyl)boronic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€88.18
250 MG
€110.23
1 G
€281.69

Vendor SKU: C63176-100MG

€88.18


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
€88.18
250 MG
€110.23
1 G
€281.69

About This Item

CAS Number:
Molecular Weight:
162.94
MDL number:

Vendor SKU: C63176-100MG

€88.18


Ships in 14 days - from Aldrich Partner.

Iupac Name

3-cyano-4-hydroxyphenylboronic acid

Inchi Code

1S/C7H6BNO3/c9-4-5-3-6(8(11)12)1-2-7(5)10/h1-3,10-12H

InChI key

PGIZXBPAULHTKH-UHFFFAOYSA-N

storage temp.

Room Temperature

Purity

95%

Country of Origin

US

Shipping Temperature

Room Temperature

Physical Form

Solid

Product Link

https://synthonix.com/63176.html

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://synthonix.com/63176.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service