Skip to Content
Merck
All Photos(1)

Key Documents

SY3H3D67C40D

Synthonix Corporation

2,4-Dichloro-6-phenyl-1,3,5-triazine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
226.06
MDL number:

Iupac Name

2,4-dichloro-6-phenyl-1,3,5-triazine

Inchi Code

1S/C9H5Cl2N3/c10-8-12-7(13-9(11)14-8)6-4-2-1-3-5-6/h1-5H

InChI key

AMEVJOWOWQPPJQ-UHFFFAOYSA-N

storage temp.

Room Temperature

Purity

98%

Country of Origin

US

Shipping Temperature

Room Temperature

Physical Form

Crystal - Powder / Colour: White - Yellow

Product Link

https://synthonix.com/50698.html

Pictograms

Exclamation mark

Signal Word

Warning

Precautionary statements

MSDS

https://synthonix.com/50698.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service