SY3H30452955
Synthonix Corporation
Methyl 6-bromo-3-chloropyrazine-2-carboxylate
Select a Size
Vendor SKU: M9913-100MG
€78.37
Select a Size
About This Item
Vendor SKU: M9913-100MG
€78.37
Iupac Name
methyl 6-bromo-3-chloro-2-pyrazinecarboxylate
Inchi Code
1S/C6H4BrClN2O2/c1-12-6(11)4-5(8)9-2-3(7)10-4/h2H,1H3
InChI key
ZDLDHAIWCZRNOT-UHFFFAOYSA-N
storage temp.
Purity
Country of Origin
Shipping Temperature
Physical Form
Product Link
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://synthonix.com/9913.html
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service