Skip to Content
Merck
All Photos(1)

Key Documents

N11404

Carbendazim

analytical standard

Synonym(s):

BCM, Methyl 2-benzimidazolecarbamate, Methyl benzimidazol-2-ylcarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9N3O2
CAS Number:
Molecular Weight:
191.19
Beilstein:
649044
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 100 mg

manufacturer/tradename

Chem Service, Inc. PS-1077

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

>300 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

COC(=O)Nc1nc2ccccc2[nH]1

InChI

1S/C9H9N3O2/c1-14-9(13)12-8-10-6-4-2-3-5-7(6)11-8/h2-5H,1H3,(H2,10,11,12,13)

InChI key

TWFZGCMQGLPBSX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Muta. 1B - Repr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Gonzalo Martínez-Fernández et al.
FEMS microbiology ecology, 91(8), fiv079-fiv079 (2015-07-18)
Study of the efficacy of methanogenesis inhibitors in the rumen has given inconsistent results, mainly due to poorly understood effects on the key microbial groups involved in pathways for methane (CH4) synthesis. The experiment described in this report was designed
Diego Forni et al.
BMC genomics, 16, 891-891 (2015-11-04)
Copy number variation (CNV) is a major component of genomic variation, yet methods to accurately type genomic CNV lag behind methods that type single nucleotide variation. High-throughput sequencing can contribute to these methods by using sequence read depth, which takes
Jae-Ran Lee et al.
Human mutation, 36(1), 69-78 (2014-09-30)
KIF1A is a neuron-specific motor protein that plays important roles in cargo transport along neurites. Recessive mutations in KIF1A were previously described in families with spastic paraparesis or sensory and autonomic neuropathy type-2. Here, we report 11 heterozygous de novo
Wojciech Dabrowski et al.
Clinical nephrology, 82(1), 41-50 (2014-06-03)
To analyze the effect and the time course of continuous veno-venous hemofiltration (CVVH) with net ultrafiltration (UF) on intra-abdominal pressure (IAP) body fluid volumes in septic shock patients with acute kidney injury (AKI). Patients were studied at baseline and after
D K Bayer et al.
Clinical and experimental immunology, 178(3), 459-469 (2014-07-22)
In areas without newborn screening for severe combined immunodeficiency (SCID), disease-defining infections may lead to diagnosis, and in some cases, may not be identified prior to the first year of life. We describe a female infant who presented with disseminated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service