Skip to Content
Merck
All Photos(1)

Key Documents

821968

TSKgel® NH2-100 (3 µm) HPLC Columns

L x I.D. 15 cm × 2 mm, amine phase

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€993.00

€993.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
€993.00

About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501

€993.00


Please contact Customer Service for Availability

Product Name

TSKgel® NH2-100 HPLC Column, phase amine, L × I.D. 15 cm × 2 mm, 3 μm particle size

material

stainless steel hardware

Agency

suitable for USP L8

product line

TSKgel®

manufacturer/tradename

Tosoh 21968

parameter

0-100% organic solvent
0.2 mL/min standard flow rate
10-50 °C temperature
200 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

15 cm × 2 mm

matrix

silica particle platform

matrix active group

amine phase

particle size

3 μm

pore size

100 Å

operating pH

2.0-7.5

separation technique

anion exchange
hydrophilic interaction (HILIC)
normal phase

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel NH2-100 amino columns expand the range of TSKgel columns for hydrophilic interaction chromatography (HILIC). Offering a different selectivity from the well known TSKgel Amide-80 columns, these novel amino-bonded phase columns stand out by providing much improved chemical stability, a prerequisite for achieving reproducible and reliable results. TSKgel NH2-100 columns are packed with 3 micron silica particles containing 100Å pores. A novel bonding strategy was adopted to improve chemical stability of the bonded phase. First the silica is reacted with a trimethylsilane endcapping reagent at a low stoichiometric ratio, before reacting residual and accessible silanol groups with trifunctional alkylaminosilane reagent. The resulting bonded phase provides a better safeguard against hydrolysis of the underlying silica. TSKgel NH2-100 columns are unique in that the bonded phase ligand not only, as expected, has a terminal primary amino group, but that the spacer also incorporates secondary as well as tertiary amino groups. Anionic compounds are retained on the column by ionic interaction. This allows for the use of gradients in salt concentration, in addition to gradient elutions with acetonitrile. Since the TSKgel NH2-100 columns have cationic sites, the columns can be used as mixed mode columns under some conditions. TSKgel NH2-100 columns are well suited for the analysis of all types of hydrophilic compounds, including carbohydrates and peptides. Due to a high ligand density and large surface area, these columns show stronger retention of polar compounds than TSKgel Amide-80 columns.

Physical form

Shipped in 85% acetonitrile - 15% water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service