Skip to Content
Merck
All Photos(1)

Key Documents

813352

TSKgel® OligoDNA RP (5 µm) HPLC Columns

L x I.D. 15 cm × 4.6 mm, C18 (octadecyl) phase

Synonym(s):

TSKgel® Reversed Phase HPLC Column

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€981.00

€981.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
€981.00

About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

€981.00


Please contact Customer Service for Availability

Product Name

TSKgel® OligoDNA RP HPLC Column, phase C18 (octadecyl), L × I.D. 15 cm × 4.6 mm, 5 μm particle size

material

PEEK

Agency

suitable for USP L1

product line

TSKgel®

manufacturer/tradename

Tosoh OligoDNA RP, 13352

extent of labeling

10% Carbon, not endcapped

parameter

no limitation salt concentration
0-100% organic solvent
0-45 °C temperature
1.2 mL/min flow rate
120 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

15 cm × 4.6 mm

matrix

silica particle platform

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

250 Å pore diameter

operating pH

2.0-7.5

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Specifically designed for purification of oligonucleotides, RNA and DNA fragments (up to 500mer), TSKgel OligoDNA RP can provide excellent separations of samples with very similar sequences. The 250Å pore size provides excellent kinetics for molecules with helix shape structures processing large radii of gyration. The 5 micron particle size provides a minumum of 7,000 plates per 15 centimeter column.

Physical form

Shipped in 50% acetonitrile - 50% water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service