Skip to Content
Merck
All Photos(1)

Key Documents

49009

Supelco

4,4′-DDD

analytical standard

Synonym(s):

1,1-Dichloro-2,2-bis(4-chlorophenyl)ethane, 4,4′-DDD, TDE

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(ClC6H4)2CHCHCl2
CAS Number:
Molecular Weight:
320.04
Beilstein:
1914072
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 1000 mg

application(s)

agriculture
environmental

SMILES string

ClC(Cl)C(c1ccc(Cl)cc1)c2ccc(Cl)cc2

InChI

1S/C14H10Cl4/c15-11-5-1-9(2-6-11)13(14(17)18)10-3-7-12(16)8-4-10/h1-8,13-14H

InChI key

AHJKRLASYNVKDZ-UHFFFAOYSA-N

Gene Information

mouse ... Esr1(13982)
rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michalina Gałgowska et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 50(11), 4125-4129 (2012-08-15)
The aim of the study was to determine the content of chlorinated hydrocarbon residues in edible mushrooms from the north-eastern part of Poland. Material consisted of two species of fungi: Xerocomus mushrooms (Xerocomus badius), Boletus mushrooms (Boletus edulis). The dried
Marko Gerić et al.
Chemosphere, 87(11), 1288-1294 (2012-02-23)
Despite that the use of DDT has been restricted for more than 40 years to malaria affected areas, low doses of this pesticide and its metabolites DDE and DDD can be found in the environment around the world. Although it
Kerry Brink et al.
Ecotoxicology and environmental safety, 79, 256-263 (2012-02-14)
The exposure and response of the catfish, Clarias gariepinus, was studied in male specimens collected in the vicinity of a DDT spraying programme to control malaria. Two sites were situated in the DDT sprayed areas and one site upstream from
Yvette Naudé et al.
Analytica chimica acta, 730, 112-119 (2012-05-29)
The organochlorine insecticide DDT (1,1,1-trichloro-2,2-bis(p-chlorophenyl)ethane) is still used for malaria vector control in certain areas of South Africa. The strict Stockholm Convention on Persistent Organic Pollutants (POPs) allows spraying on the inside of traditional dwellings with DDT. In rural villages
Mehmet Isleyen et al.
Journal of agricultural and food chemistry, 60(4), 1113-1121 (2012-01-10)
The grafting of melon plants onto cucurbit rootstocks is a common commercial practice in many parts of the world. However, certain cucurbits have been shown to accumulate large quantities of weathered persistent organic pollutants from the soil, and the potential

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service