Skip to Content
Merck
All Photos(1)

Key Documents

48706

Supelco

Aroclor 1242 solution

certified reference material, 200 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

Agency

EPA 505,508,8082
MISA (Canada) Group 27

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

200 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-30°C

InChI

1S/C12H6Cl4/c13-7-1-3-9(11(15)5-7)10-4-2-8(14)6-12(10)16/h1-6H

InChI key

QORAVNMWUNPXAO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aroclor 1242 is a mixture of polychlorinated biphenyls, which can find wide applications as a dielectric medium in capacitors and transformers.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT RE 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup - Solvent

Flash Point(C)

11 °C - closed cup - Solvent

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Degradation of polychlorinated biphenyl mixtures (Aroclors 1242, 1254, and 1260) by the white rot fungus Phanerochaete chrysosporium as evidenced by congener-specific analysis
Yadhav JS, et al.
Applied and Environmental Microbiology, 61(7), 2560-2565 (1995)
Use and health effects of Aroclor 1242, a polychlorinated biphenyl, in an electrical industry
Ouw HK, et al.
Archives of Environmental Health, 31(4), 189-194 (1976)
Tarek Saba et al.
Chemosphere, 82(9), 1321-1328 (2010-12-31)
Aroclor 1242 contains a high percentage of lightly chlorinated congeners, which makes it susceptible to congener profile alterations as a result of physical-chemical environmental weathering by water washing, evaporation, and volatilization. The analysis of the variability of congener profiles in
Jorge L M Rodrigues et al.
Applied and environmental microbiology, 72(4), 2476-2482 (2006-04-07)
Burkholderia xenovorans strain LB400, which possesses the biphenyl pathway, was engineered to contain the oxygenolytic ortho dehalogenation (ohb) operon, allowing it to grow on 2-chlorobenzoate and to completely mineralize 2-chlorobiphenyl. A two-stage anaerobic/aerobic biotreatment process for Aroclor 1242-contaminated sediment was
Lars Rehmann et al.
Biotechnology and bioengineering, 99(5), 1273-1280 (2007-10-12)
This article demonstrates the feasibility of a novel process concept for the remediation of PCB contaminated soil. The proposed process consists of PCB extraction from soil using solid polymer beads, followed by biodegradation of the extracted PCBs in a solid-liquid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service