Skip to Content
Merck
All Photos(6)

Key Documents

ZRB2251

Sigma-Aldrich

Anti-Siglec1/CD169 Antibody, clone 1O8 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

CD169, Sialic acid-binding Ig-like lectin 1, Sialic acid-binding immunoglobulin-like lectin 1, Sialoadhesin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1O8, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

Internal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... SIGLEC1(6614)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1O8 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Sialic acid-binding immunoglobulin-like lectin 1 (Siglec1/CD169). It targets an epitope within 21 amino acids from the internal region.

Immunogen

KLH-conjugated linear peptide corresponding to 21 amino acids from the internal region of human Sialic acid-binding immunoglobulin-like lectin 1 (Siglec1/CD169).

Application

Quality Control Testing

Evaluated by Western Blotting in Human Lymph Node tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Siglec1/CD169 in Human Lymph Node tissue lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Siglec1/CD169 in lysate from SH-SY5Y cells and Human tonsil tissue.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Siglec1/CD169 in Human tonsil tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound Siglec1/CD169 peptide with a KD of 9.8 x 10-8 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Sialoadhesin (UniProt Q9BZZ2; also known as CD169, Sialic acid-binding Ig-like lectin 1, Sialic acid-binding immunoglobulin-like lectin 1, Siglec-1) is encoded by the SIGLEC1 (also known as SN) gene (Gene ID 6614) in human. Sialoadhesin/CD169/Siglec1 is the founding member of the sialic acid-binding immunoglobulin lectins (Siglec) expressed on a variety of leukocytes and stromal cells. Siglec1 is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-19), which is subsequently cleaved off to produce the mature for that contains a large extracellular domain (aa 20-1641), a transmembrane domain (aa 1642-1662), and a short cytoplasmic domain (aa 1663-1709) with no known signal transduction or endocytosis motifs. The extracellular portion contains an N-terminal Ig-like V-type domain (aa 20-136), followed by sixteen Ig-like C2-type domains. Siglec1 is highly expressed on the subcapsular sinus (SCS) and medullary macrophages in lymph node (LN) and on marginal metallophilic macrophages in the marginal zone (MZ) of spleen. Although sialic acids are found on the surface of all cells and most secreted proteins, only heavily sialylated, multimeric structures are captured by Siglec1 on macrophages via its N-terminal Ig-like V-type domain due to a low (millimolar) Affinity toward sialylic acid. Siglec1-mediated sialic-acid recognition allows macrophages to engage sialylated pathogens and stimulate inflammatory responses. Siglec1 is also reported to capture of B cell-derived exosomes via their surface-expressed α2, 3-linked sialic acids. In the lymph node, exosomes are not retained in the subcapsular sinus of Siglec1-knockout mice and penetrate deeper into the paracortex, causing an enhanced response to antigen-pulsed exosomes among Siglec1-/- mice. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, Affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Saunderson, SC., et al. (2014). Blood. 123(2); 208-216; O′Neil. OSG., et al. (2013). Immunology. 138(3); 198-207; Jones, C., et al. (2003). Mol. Microbiol. 49(5); 1213-1225).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

Affinity is a trademark of Mine Safety Appliances Co.
ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service