Skip to Content
Merck
All Photos(1)

Key Documents

Z744323

Wilmad® Suprasil® Electrolytic Assembley

for use with Varian, Radiopan, pack of 1

Synonym(s):

EPR kinetic study assembly, electrolytic cell assembly

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115600
NACRES:
NB.15

packaging

pack of 1

manufacturer/tradename

Wilmad WG-810-Q

compatibility

for use with Varian, Radiopan

Looking for similar products? Visit Product Comparison Guide

General description

Electrochemistry is employed in many EPR kinetic studies to generate free radicals and avoids alkali metal reductions, the associated sample sensitivity to air or moisture and background signal. A complete cell includes two Calomel Reference Electrodes, a pair of Platinum Electrodes, and a gas bubbler for deoxygenation. The Electrolytic Cell Assembly comes as a complete unit with CFQ or ′Suprasil′ Flat Cell. The Flat Electrolytic Cells have two Standard Taper 10/30 outer joints at the top, one for the Gas Bubbler and a Platinum Electrode and at the botom a single Standard Taper 5/20 outer joint for the other Platinum Electrode, the remainder of the Flat Cell is CFQ. A minimum 15mL of sample solution needs to make contact with the electrodes and gas bubbler. The cell width for JEOL spectrometers is 10mm outside (WG-810-J-Q), for other spectrometers, the cell width is 10.5mm outside (WG-810-Q). The Gas Bubbler Assembly is a three-piece Pyrex® chamber which holds part of your sample it has three Standard Taper 10/30 outer joints at the top for the Gas Bubbler and two Calomel Electrodes. The lower Standard Taper 10/30 inner joint fits into the top of the Flat Electrolytic Cell. Nitrogen should be bubbled through the solution before and during the experiment to remove paramagnetic oxygen and mix the sample during electrolysis. The Platinum Electrodes are prepared using 0.016? (0.4mm) diameter wire. The upper electrode, WG-810-C, uses 8? (203mm) of platinum and acts as the working electrode in oxidation reactions. The lower electrode, WG-810-D, has only 2? (50mm) of platinum and acts as the auxiliary electrode in oxidation reactions. If you add a few drops of mercury to the cell, the lower electrode can be used as the working cathode in reduction reactions. Each Platinum Electrode incorporates a Pt-to-W weld. The tungsten (W) external wire lets you quickly clip your potentiostat leads to the electrode.Electrolysis Limits in Water (vs. Saturated Calomel Electrode)

Application

EPR

Features and Benefits

  • Complete cell includes two Calomel Reference Electrodes, a pair of Platinum Electrodes, and a gas bubbler
  • Electrolytic Cell Assembly comes as a complete unit with CFQ or Suprasil® Flat Cell

Legal Information

PYREX is a registered trademark of Corning, Inc.
Suprasil is a registered trademark of HERAEUS QUARZGLAS GMBH & CO.
Wilmad is a registered trademark of SP Industries

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service