Skip to Content
Merck
All Photos(3)

Documents

WH0010766M1

Sigma-Aldrich

Monoclonal Anti-TOB2 antibody produced in mouse

clone 2F2-1A7, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-TOB4, Anti-TOBL, Anti-TROB2, Anti-transducer of ERBB2, 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2F2-1A7, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2bκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TOB2(10766)

General description

TOB2 belongs to the TOB (see TOB1; MIM 605523)/BTG1 (MIM 109580) family of antiproliferative proteins, which are involved in the regulation of cell cycle progression.[supplied by OMIM

Immunogen

TOB2 (AAH38957, 1 a.a. ~ 344 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MQLEIKVALNFIISYLYNKLPRRRADLFGEELERLLKKKYEGHWYPEKPLKGSGFRCVHIGEMVDPVVELAAKRSGLAVEDVRANVPEELSVWIDPFEVSYQIGEKGAVKVLYLDDSEGCGAPELDKEIKSSFNPDAQVFVPIGSQDSSLSNSPSPSFGQSPSPTFIPRSAQPITFTTASFAATKFGSTKMKKGGGAASGGGVASSGAGGQQPPQQPRMARSPTNSLLKHKSLSLSMHSLNFITANPAPQSQLSPNAKEFVYNGGGSPSLFFDAADGQGSGTPGPFGGSGAGTCNSSSFDMAQVFGGGANSLFLEKTPFVEGLSYNLNTMQYPSQQFQPVVLAN

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Feng et al.
Oncogene, 30(19), 2242-2251 (2011-01-19)
The c-Myc transcription factor activates a cascade of downstream targets to form a complex transcriptional program that ultimately leads to cellular transformation. Although a large number of protein-encoding genes as well as non-coding RNAs were identified as Myc targets, only

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service