Skip to Content
Merck
All Photos(1)

Documents

SRP5135

Sigma-Aldrich

SOD1, GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

ALS, ALS1, Cu/Zn-SOD, Homodimer, IPOA, SOD

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~45 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SOD1(6647)

General description

SOD1 (superoxide dismutase 1) is the major soluble cytoplasmic enzyme responsible for destroying harmful free superoxide radicals in the body thereby providing defence against oxygen free-radical toxicity. Soluble cytoplasmic SOD1 is a copper- and zinc-containing enzyme and the SOD1 gene maps to chromosome 21q22. Mutations in the SOD1 gene have been implicated to be the cause of familial amyotrophic lateral sclerosis, increased age-related muscle mass loss, early development of cataracts, macular degeneration, thymic involution, hepatocellular carcinoma, and shortned lifespan.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Al-Chalabi et al.
Current opinion in neurology, 13(4), 397-405 (2000-09-02)
The mechanisms by which mutations of the SOD1 gene cause selective motor neuron death remain uncertain, although interest continues to focus on the role of peroxynitrite, altered peroxidase activity of mutant SOD1, changes in intracellular copper homeostasis, protein aggregation, and
L Sherman et al.
Proceedings of the National Academy of Sciences of the United States of America, 80(18), 5465-5469 (1983-09-01)
Cytoplasmic superoxide dismutase (SOD-1; EC 1.15.1.1) is encoded by human chromosome 21. The SOD-1 gene locus is located at chromosomal region 21q22, which is involved in Down syndrome. cDNA clones containing sequences of human SOD-1 were previously isolated. In the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service