Skip to Content
Merck
All Photos(2)

Key Documents

SRP0134

Sigma-Aldrich

EZH2/EED/SUZ12/RbAp48/AEBP2 human

recombinant, expressed in baculovirus infected insect cells

Synonym(s):

ENX-1, EZH2, Enhancer of zeste homolog 2, KMT6A, Lysine N-methyltransferase 6A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected insect cells

Assay

≥80% (SDS-PAGE)

form

aqueous solution

mol wt

48 kDa (RbAp48)
51 kDa (EED)
53 kDa (AEBP2)
86 kDa (EZH2)
87 kDa (SUZ12)

packaging

pkg of 50 μg

concentration

>0.02 mg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... EZH2(2146)

General description

EZH2, EED, SUZ12, RbAp48 and AEBP2 are components of the polycomb repressive complex 2 (PRC2). The highly conserved complex methylates lysine 27 of histone H3. It also has a role in gene silencing and transcription repression.

Application

EZH2/EED/SUZ12/RbAp48/AEBP2 human is useful for the study of enzyme kinetics, screening inhibitors and selectivity profiling.

Unit Definition

One unit is defined as the amount of enzyme required to methylate 1 pmol of substrate/min at 37°C.

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 138 mM NaCl, 0.05% Tween-20 and 10% glycerol.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Astemizole Arrests the Proliferation of Cancer Cells by Disrupting the
EZH2-EED Interaction of Polycomb Repressive Complex 2
Xiangqian Kong
Journal of Medicinal Chemistry (2014)
The Polycomb Complex PRC2 and its Mark in Life
Raphael Margueron
Nature, 469(7330), 343-349 (2011)
Molecular architecture of human polycomb repressive complex 2
Claudio Ciferre
eLife, 1, e00005-e00005 (2012)
Decoding protein methylation function with thermal stability analysis.
Sayago, et al.
Nature Communications, 14, 3016-3016 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service