Skip to Content
Merck
All Photos(2)

Key Documents

SAB5300417

Sigma-Aldrich

Monoclonal Anti-MTHFR antibody produced in mouse

clone 5D3, ascites fluid

Synonym(s):

MTHFR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

5D3, monoclonal

mol wt

75 kDa

species reactivity

human, rat

technique(s)

direct ELISA: 1:10,000
immunohistochemistry: 1:200-1:1,000
western blot: 1:500-1:2,000

isotype

IgG1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MTHFR(4524)

Immunogen

Purified recombinant fragment of human MTHFR expressed in E.coli.
Mouse monoclonal antibody raised against MTHFR

Physical form

Ascitic fluid containing 0.03% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuxiang Zheng et al.
Scientific reports, 9(1), 4190-4190 (2019-03-14)
Methylenetetrahydrofolate reductase (MTHFR) catalyzes the irreversible conversion of 5,10-methylene-tetrahydrofolate (THF) to 5-methyl-THF, thereby committing one-carbon units to the methionine cycle. While MTHFR has long been known to be allosterically inhibited by S-adenosylmethionine (SAM), only relatively recently has N-terminal multisite phosphorylation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service