SAB5201720
Monoclonal Anti-Hsf1 - Atto 594 antibody produced in rat
clone 4B4, purified immunoglobulin
Synonym(s):
Anti-HSTF1
About This Item
biological source
rat
Quality Level
conjugate
Atto 594 conjugate
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
4B4, monoclonal
form
buffered aqueous solution
mol wt
antigen predicted mol wt 85 kDa
species reactivity
human, bovine, mouse, rat, guinea pig, rabbit, monkey, hamster
concentration
1 mg/mL
technique(s)
ELISA: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable
isotype
IgG1
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
mouse ... Hsf1(15499)
Immunogen
Features and Benefits
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service