Skip to Content
Merck
All Photos(5)

Key Documents

SAB4504122

Sigma-Aldrich

Anti-phospho-CaMK4 (pThr196/200) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€390.00

€390.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€390.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€390.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 51 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr196/pThr200)

Gene Information

human ... CAMK4(814)

General description

Calcium/calmodulin-dependent protein kinase IV (CAMK4) is a multifunctional serine/threonine protein kinase. It is present in cells of the nervous and immune systems. This gene is located on human chromosome 5q22.

Immunogen

The antiserum was produced against synthesized peptide derived from human CaMK4 around the phosphorylation site of Thr196/200.

Immunogen Range: 166-215

Biochem/physiol Actions

Calcium/calmodulin-dependent kinase 4 (CAMK4) controls blood pressure.This protein plays a major role in the spermatogenesis by phosphorylating protamines.. It controls vascular tone. Absence of CaMK4 gene promotes hypertension.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CaMK4 Gene Deletion Induces Hypertension
Santulli G
Journal of the American Heart Association (2012)
CAMK4 gene variation is associated with hypertension in a Uygur population
Chen SZ, et al.
Genetics and molecular research : GMR, 15(1) (2016)
Calcium/calmodulin-dependent kinase IV in immune and inflammatory responses: novel routes for an ancient traveller
Racioppi L and Means AR
Trends in Immunology, 29(12), 600-607 (2008)
Akanksha Roy et al.
International journal of molecular sciences, 25(6) (2024-03-28)
Inherited retinal degenerative diseases (IRDs) are a group of rare diseases that lead to a progressive loss of photoreceptor cells and, ultimately, blindness. The overactivation of cGMP-dependent protein kinase G (PKG), one of the key effectors of cGMP-signaling, was previously
A Khattri et al.
International journal of andrology, 35(6), 810-818 (2012-08-18)
Calcium/calmodulin-dependent protein kinase IV (CAMK4) is a multifunctional serine/threonine protein kinase, which plays an important role in the spermatogenesis by phosphorylating protamines. It has been shown to be involved in the regulation of human sperm motility. Moreover, the Camk4 knockout

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service