Skip to Content
Merck
All Photos(2)

Key Documents

SAB4501345

Sigma-Aldrich

Anti-SENP1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

SENP1, Sentrin-specific protease 1, Sentrin/SUMO-specific protease SENP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 73 kDa

species reactivity

human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SENP1(29843)

Related Categories

General description

Anti-SENP1 Antibody detects endogenous levels of total SENP1 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human SENP1.

Immunogen Range: 1-50

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yunfei Liu et al.
Current molecular medicine, 18(8), 523-532 (2019-01-15)
It is now well established that protein sumoylation acts as an important regulatory mechanism modulating functions over three thousand proteins. In the vision system, protein conjugation with SUMO peptides can regulate differentiation of multiple ocular tissues. Such regulation is often
Jia-Wen Xiang et al.
Current molecular medicine, 19(1), 48-53 (2019-03-12)
It has been well established that sumoylation acts as an important regulatory mechanism that controls many different cellular processes. We and others have shown that sumoylation plays an indispensable role during mouse eye development. Whether sumoylation is implicated in ocular
Fang-Yuan Liu et al.
Aging cell, 19(10), e13222-e13222 (2020-08-23)
Sumoylation is one of the key regulatory mechanisms in eukaryotes. Our previous studies reveal that sumoylation plays indispensable roles during lens differentiation (Yan et al. 2010. Proc Natl Acad Sci USA. 107:21034-21039; Gong et al. 2014. Proc Natl Acad Sci
Jia-Wen Xiang et al.
Current molecular medicine, 18(8), 533-541 (2019-01-15)
Protein Sumoylation is one of the most important and prevalent posttranscriptional modification. Increasing evidence have shown that the SENPs (sentrin/SUMOspecific proteases) are critical for steady-state levels of SUMO modification of target proteins, and protein de-sumoylation modulates a great diversity of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service