Skip to Content
Merck
All Photos(1)

Key Documents

SAB1412171

Sigma-Aldrich

ANTI-SUPT16H antibody produced in mouse

clone 3G9, purified immunoglobulin, buffered aqueous solution

Synonym(s):

CDC68, FACT, FACTP140, FLJ10857, SUPT16H

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€491.00

€491.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€491.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€491.00


Please contact Customer Service for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3G9, monoclonal

form

buffered aqueous solution

mol wt

antigen 37.62 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SUPT16H(11198)

General description

Transcription of protein-coding genes can be reconstituted on naked DNA with only the general transcription factors and RNA polymerase II. However, this minimal system cannot transcribe DNA packaged into chromatin, indicating that accessory factors may facilitate access to DNA. One such factor, FACT (facilitates chromatin transcription), interacts specifically with histones H2A/H2B to effect nucleosome disassembly and transcription elongation. FACT is composed of an 80 kDa subunit and a 140 kDa subunit; this gene encodes the 140 kDa subunit. (provided by RefSeq)

Immunogen

SUPT16H (NP_009123, 608 a.a. ~ 715 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
PGEQTVPALNLQNAFRIIKEVQKRYKTREAEEKEKEGIVKQDSLVINLNRSNPKLKDLYIRPNIAQKRMQGSLEAHVNGFRFTSVRGDKVDILYNNIKHALFQPCDGE

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service