Skip to Content
Merck
All Photos(2)

Key Documents

SAB1408617

Sigma-Aldrich

Anti-PFKFB1 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

F6PK, HL2K, MGC116715, MGC116717, PFRX

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€489.00

€489.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€489.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€489.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~54.7 kDa

species reactivity

human

technique(s)

immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PFKFB1(5207)

General description

6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 1 (PFKFB1) is encoded by the gene mapped to human chromosome Xp11.21. The encoded protein is one among the four isoenzymes of 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase. PFKFB1 is expressed predominantly in liver.
This gene encodes a member of the family of bifunctional 6-phosphofructo-2-kinase:fructose-2,6-biphosphatase enzymes. The enzyme forms a homodimer that catalyzes both the synthesis and degradation of fructose-2,6-biphosphate using independent catalytic domains. Fructose-2,6-biphosphate is an activator of the glycolysis pathway and an inhibitor of the gluconeogenesis pathway. Consequently, regulating fructose-2,6-biphosphate levels through the activity of this enzyme is thought to regulate glucose homeostasis. (provided by RefSeq)

Immunogen

PFKFB1 (NP_002616.1, 1 a.a. ~ 471 a.a) full-length human protein.

Sequence
MSPEMGELTQTRLQKIWIPHSSGSSRLQRRRGSSIPQFTNSPTMVIMVGLPARGKTYISTKLTRYLNWIGTPTKVFNLGQYRREAVSYKNYEFFLPDNMEALQIRKQCALAALKDVHNYLSHEEGHVAVFDATNTTRERRSLILQFAKEHGYKVFFIESICNDPGIIAENIRQVKLGSPDYIDCDREKVLEDFLKRIECYEVNYQPLDEELDSHLSYIKIFDVGTRYMVNRVQDHIQSRTVYYLMNIHVTPRSIYLCRHGESELNIRGRIGGDSGLSVRGKQYAYALANFIQSQGISSLKVWTSRMKRTIQTAEALGVPYEQWKALNEIDAGVCEEMTYEEIQEHYPEEFALRDQDKYRYRYPKGESYEDLVQRLEPVIMELERQENVLVICHQAVMRCLLAYFLDKSSDELPYLKCPLHTVLKLTPVAYGCKVESIYLNVEAVNTHREKPENVDITREPEEALDTVPAHY

Application

Anti-PFKFB1 antibody produced in rabbit has been used in western blotting.

Biochem/physiol Actions

6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 1 (PFKFB1) catalyzes the synthesis and degradation of fructose-2,6-bisphosphate. It plays a vital role in regulation of both glycolysis and gluconeogenesis. PFKFB1 also facilitates cell growth and apoptosis.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gene expression analysis of epidermolysis bullosa simplex with mottled pigmentation
Bchetnia M, et al.
Journal of Dermatological Science, 69(1), 80-82 (2013)
6-Phosphofructo-2-kinase/fructose-2, 6-biphosphatase 4 is essential for p53-null cancer cells.
Ros S, et al.
Oncogene, 36(23), 3287-3287 (2017)
A pericentric inversion of chromosome X disrupting F8 and resulting in haemophilia A
Xin Y, et al.
Journal of Clinical Pathology (2017)
R S Batra et al.
Genomics, 40(2), 358-361 (1997-03-01)
6-Phosphofructo-2-kinase/fructose-2,6-bisphosphatase (PFK-2/FBPase-2) catalyzes the synthesis and degradation of fructose-2,6-bisphosphate, a potent regulator of glycolysis. Previous studies assigned the gene for human liver PFK-2/FBPase-2 (HGMW-approved symbol PFKFB1) to the X chromosome; however, precise localization remained ambiguous, with the gene variously placed
S Ros et al.
Oncogene, 36(23), 3287-3299 (2017-01-17)
The bifunctional enzyme 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase-4 (PFKFB4) controls metabolic flux through allosteric regulation of glycolysis. Here we show that p53 regulates the expression of PFKFB4 and that p53-deficient cancer cells are highly dependent on the function of this enzyme. We found that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service