Skip to Content
Merck
All Photos(5)

Key Documents

SAB1407188

Sigma-Aldrich

Anti-FBXO28 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

FLJ10766, Fbx28, KIAA0483

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~41.1 kDa

species reactivity

mouse, human, rat

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FBXO28(23219)

General description

Members of the F-box protein family, such as FBXO28, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).[supplied by OMIM

Immunogen

FBXO28 (NP_055991.1, 1 a.a. ~ 368 a.a) full-length human protein.

Sequence
MAAAAEERMAEEGGGGQGDGGSSLASGSTQRQPPPPAPQHPQPGSQALPAPALAPDQLPQNNTLVALPIVAIENILSFMSYDEISQLRLVCKRMDLVCQRMLNQGFLKVERYHNLCQKQVKAQLPRRESERRNHSLARHADILAAVETRLSLLNMTFMKYVDSNLCCFIPGKVIDEIYRVLRYVNSTRAPQRAHEVLQELRDISSMAMEYFDEKIVPILKRKLPGSDVSGRLMGSPPVPGPSAALTTMQLFSKQNPSRQEVTKLQQQVKTNGAGVTVLRREISELRTKVQEQQKQLQDQDQKLLEQTQIIGEQNARLAELERKLREVMESAVGNSSGSGQNEESPRKRKKATEAIDSLRKSKRLRNRK

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service