Skip to Content
Merck
All Photos(2)

Key Documents

SAB1400205

Sigma-Aldrich

Anti-PGM5 antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-PGMRP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PGM5(5239)

Related Categories

General description

Phosphoglucomutases (EC 5.2.2.2.), such as PGM5, are phosphotransferases involved in interconversion of glucose-1-phosphate and glucose-6-phosphate. PGM activity is essential in formation of carbohydrates from glucose-6-phosphate and in formation of glucose-6-phosphate from galactose and glycogen (Edwards et al., 1995 [PubMed 8586438]).[supplied by OMIM

Immunogen

PGM5 (ENSP00000366540, 1 a.a. ~ 506 a.a) full-length human protein.

Sequence
MVVGSDGRYFSRTAIEIVVQMAAANGIGRLIIGQNGILSTPAVSCIIRKIKAAGGIILTASHCPGGPGGEFGVKFNVANGGPAPDVVSDKIYQISKTIEEYAICPDLRIDLSRLGRQEFDLENKFKPFRVEIVDPVDIYLNLLRTIFDFHAIKGLLTGPSQLKIRIDAMHGVMGPYVRKVLCDELGAPANSAINCVPLEDFGGQHPDPNLTYATTLLEAMKGGEYGFGAAFDADGDRYMILGQNGFFVSPSDSLAIIAANLSCIPYFRQMGVRGFGRSMPTSMALDRVAKSMKVPVYETPAGWRFFSNLMDSGRCNLCGEESFGTGSDHLREKDGLWAVLVWLSIIAARKQSVEEIVRDHWAKFGRHYYCRFDYEGLDPKTTYYIMRDLEALVTDKSFIGQQFAVGSHVYSVAKTDSFEYVDPVDGTVTKKQGLRIIFSDASRLIFRLSSSSGVRATLRLYAESYERDPSGHDQEPQAVLSPLIAIALKISQIHERTGRRGPTVIT

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service