Skip to Content
Merck
All Photos(1)

Key Documents

RAB0414

Sigma-Aldrich

Retinol Binding Protein 4 EIA Kit

for serum, plasma, culture supernatant and cell lysates

Synonym(s):

RBP4

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€500.00

€500.00


Please contact Customer Service for Availability


Select a Size

Change View
1 KIT
€500.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€500.00


Please contact Customer Service for Availability

species reactivity

human, mouse, rat

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
enzyme immunoassay: suitable

input

sample type serum
sample type plasma
sample type cell lysate
sample type culture supernatant(s)

assay range

inter-assay cv: <15%
intra-assay cv: <10%
sensitivity: 460 pg/mL
standard curve range: 0.1-1000 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... RBP4(5950)

General description

The RBP4 Enzyme Immunoassay (EIA) Kit is an in vitro quantitative assay for detecting RBP4 peptide based on the principle of Competitive Enzyme Immunoassay. EIA-RBP-1 detects RBP4 (183aa). It detects all other active forms including RBP4-182,181,179,176.

Immunogen

Retinol Binding Protein 4 (RBP4), synthetic peptide

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Kit Components Also Available Separately

Product No.
Description
SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service